DuPont™ Tychem® 5000 Bib Coverall and Jacket Combo
Supplier: DuPont
C3750TTN2X000600 C3750TTN3X000600 C3750TTN4X000600 C3750TTNLG000600 C3750TTNMD000600 C3750TTNSM000600 C3750TTNXL000600
76187-458CS 728.3 USD InStock
76187-458 76187-460 76187-462 76187-464 76187-466 76187-468 76187-470
DuPont™ Tychem® 5000 Bib Coverall and Jacket Combo
Disposable Garments
Tychem® 5000 fabric is composed of a multi-layer film barrier laminated to a durable 2.0 oz/yd2 polypropylene substrate.
  • Coverall and jacket combo
  • Jam fit cuffs
  • Open ankles
  • Storm flap on jacket
  • Adjustable webbing straps with buckle closure on coveralls
  • Taped seams
Tychem® 5000 fabric is strong and durable for rigorous activities and rugged situations involving liquid splash and provides barrier to a broad range of chemicals. Typical Applications: chemical handling, petro-chemical, hazardous materials/waste clean-up, fire departments, industrial hazmat teams, utilities, and domestic preparedness. Commonly used in domestic preparedness for situations where the potential to exposure to chemicals exist. These coveralls and jackets are tan.
This ensemble consists of a bib-style coverall and jacket combination. Components are also available separately. This jacket comes with a jam fit cuff for customer-installed jam fit glove systems.
Certificaciones: Thickness meets ASTM D1777.
Order Now
ORDER
SPECIFICATIONS
PRODUCTOS COMPLEMENTARIOS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty