Plastic Vials, Chemglass
Supplier: Chemglass
CV-1007-1232 CV-1008-1232 CV-1039-1232
75819-582PK 29.63 USD InStock
75819-582 75819-584 75819-640
Plastic Vials, Chemglass
Vials
These plastic vials are used when glass vails cause problems in chromatographic reproducibility.
  • Vials are as inert as glass and have a hydrophobic surface
  • Made of PP
  • 9 mm thread
PP vials can be used with Proteins, DNA and bioplymers.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty