DuPont™ Tychem® 6000 Coveralls with Respirator Hood and Attached Socks
Supplier: DuPont
TF169TGY2X000600 TF169TGY2X0006TV TF169TGYMD000600 TF169TGYMD0006TV TF169TGYSM000600 TF169TGYSM0006TV TF169TGY3X000600 TF169TGY3X0006TV TF169TGY4X000600 TF169TGY4X0006TV TF169TGY5X000600 TF169TGY5X0006TV TF169TGYLG000600 TF169TGYLG0006TV TF169TGYXL000600 TF169TGYXL0006TV
76187-654CS 472.41 USD InStock
76187-654 76187-656 76187-658 76187-660 76187-662 76187-664 76187-842 76187-844 76187-846 76187-848 76187-850 76187-852 76187-854 76187-856 76187-858 76187-860
DuPont™ Tychem® 6000 Coveralls with Respirator Hood and Attached Socks
Disposable Garments
Tychem® 6000 garments are made of a proprietary barrier film laminated to a Tyvek® substrate.
  • Respirator fit hood
  • Elastic wrists
  • Attached socks
  • Storm flap
  • Taped seams
The fabric provides at least 30 minutes of protection against over 181 challenge chemicals including chemical warfare agents and toxic industrial chemicals. Tychem® 6000 garments are strong, durable and lightweight, and they are available in both high-visibility orange and low-visibility gray fabrics, making them a preferred choice for law enforcement, emergency medical services (EMS) technicians and military personnel.
Taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing. Three-piece hood design with longer zipper that extends to chin allows for complete coverage of neck area and good fit around a respirator. Storm flap covers zipper, which can be sealed by the wearer with adhesive strip to prevent intrusion at zipper. Elastic openings at wrists allow for a tighter fit. Integrated socks are composed of the same garment material.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty