Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

FMOC isothiocyanate ≥95% (by NMR), pale yellow solid

Supplier: A.W. SCHÜRR GmbH

12619-25G 12619-1G 12619-5G
102562-954EA 2417.94 USD InStock
102562-954 102567-020 102630-156
FMOC isothiocyanate ≥95% (by NMR), pale yellow solid
FMOC isothiocyanate
Synonym: Fmoc-NCS


Formula: C₁₆H₁₁NO₂S
Melting Pt: 36…49 °C
Storage Temperature: Freezer
MDL Number: MFCD01861169
CAS Number: 199915-38-3
Order Now
Specification Test Results
Infrared Spectrum Conforms to Structure
13C NMR Conforms to Structure
1H NMR Conforms to Structure
Purity ≥95% (NMR)

 

ORDER

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty