Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tetrakis[(S)-(-)-N-(p-dodecylphenylsulfonyl)prolinato]dirhodium(II) Rh2(S-DOSP)4, green powder

Warning

45-2101-250MG 45-2101-50MG
100198-812EA 266.09 USD InStock
100198-812 100198-810
Tetrakis[(S)-(-)-N-(p-dodecylphenylsulfonyl)prolinato]dirhodium(II) Rh2(S-DOSP)4, green powder
Tetrakis[(S)-(-)-N-(p-dodecylphenylsulfonyl)prolinato]dirhodium(II) Rh2(S-DOSP)4


Formula: C₉₂H₁₄₄N₄O₁₆Rh₂S₄
MW: 1896.22 g/mol
MDL Number: MFCD04974246
CAS Number: 179162-34-6
Order Now
Specification Test Results
Spec. rotation —165° (c 1.0, CHCl₃)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty