GRK5 Kinase Enzyme System, Promega
Supplier: EBRO Electronic GmbH
V3981 V3982
PAV3981EA 656.55 USD InStock
PAV3981 PAV3982
GRK5 Kinase Enzyme System, Promega
Assays Enzyme Assays
Recombinant full-length human GRK5 was expressed by baculovirus in Sf9 cells using an N-terminal GST tag. GRK5 is a member of the guanine nucleotide-binding protein (G protein)-coupled receptor kinase subfamily of the Ser/Thr protein kinase family.
  • Easily Screen and Profile GRK5 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Recombinant full-length human GRK5 was expressed by baculovirus in Sf9 cells using an N-terminal GST tag. GRK5 is a member of the guanine nucleotide-binding protein (G protein)-coupled receptor kinase subfamily of the Ser/Thr protein kinase family. It phosphorylates the activated forms of G protein-coupled receptors thus initiating their deactivation. GRK5 plays a role in regulating the motility of polymorphonuclear leukocytes (PMNs). Desensitization of G protein-coupled receptors regulates the number of polymorphonuclear leukocytes (PMNs), as well as their motility and ability to stop pathogens or target cells upon contact, and this desensitization is mediated by GRK5. ADP-Glo Kinase Assay is a luminescent kinase assay that measures ADP formed from a kinase reaction; ADP is converted into ATP, which is a substrate in a reaction catalyzed by Ultra-Glo Luciferase that produces light. The luminescent signal positively correlates with ADP amount and kinase activity. The assay is well suited for measuring the effects chemical compounds have on the activity of a broad range of purified kinases, making it ideal for both primary screening as well as kinase selectivity profiling. The ADP-Glo Kinase Assay can be used to monitor the activity of virtually any ADP-generating enzyme (e.g., kinase or ATPase) using up to 1mM ATP. Kinase Enzyme System contains: Kinase: GRK5, 10ug (Human, recombinant full-length). MW: ~95kDa. Substrate: Native casein protein purified from bovine milk. Other: Reaction Buffer, DTT. GRK5 NCBI Database Entry: www.ncbi.nlm.nih.gov/gene/2869/. Visit www.promega.com/kinase/ to see all Kinase Enzyme Systems.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty