Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Alpelisib free base ≥99% (by HPLC)

Supplier: SAFE AND SURE NEDERLAND B.V.

CHM214-0025 CHM214-0050 CHM214-0100 CHM214-0200 CHM214-0500 CHM214-1000 CHM214-0005
76203-090EA 316 USD InStock
76203-090 76203-098 76203-100 76203-102 76203-104 76203-106 76203-506
Alpelisib free base ≥99% (by HPLC)
Alpelisib
Alpelisib is a potent and selective PI3Kα inhibitor with IC50 of 5 nM in a cell-free assay, and minimal effect on PI3Kβ/γ/δ

  • Potent and selective PI3Kα inhibitor

White solid.

Soluble in DMSO.


Formula: C₁₉H₂₂F₃N₅O₂S
MW: 441.47 g/mol
Storage Temperature: Freezer
MDL Number: MFCD22417085
CAS Number: 1217486-61-7
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty