Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Binimetinib free base ≥99% (by HPLC)


CHM215-0050 CHM215-0100 CHM215-0200 CHM215-1000
76203-110EA 175 USD InStock
76203-110 76203-112 76203-114 76203-116 76203-118
Binimetinib free base ≥99% (by HPLC)
Binimetinib


Formula: C₁₇H₁₅BrF₂N₄O₃
Storage Temperature: Freezer
MDL Number: MFCD22124525
CAS Number: 606143-89-9
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty