Human Recombinant TGF-A (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC.
100-209-25UG 100-209-100UG 100-209-250UG 100-209-500UG 100-209-1MG
76199-352EA 83.2 USD InStock
76199-352 76199-354 76199-356 76199-358 76199-360
Human Recombinant TGF-A (from E. coli)
Proteins and Peptides
Tumor growth factor alpha (TGF-α) is a member of the epidermal growth factor (EGF) family. TGF-α function is mediated through binding the EGF receptor (EGFR) to activate receptor tyrosine kinase signaling. TGF-α functions as a mitogen to activate epithelial cell proliferation, growth, and differentiation.
  • High quality
  • Low endotoxin
  • Affordable
In the gastric mucosa, TGF-α production inhibits gastric acid secretion and therefore plays a central role in the pathogenesis of the stomach syndrome Ménétrier's disease. TGF-α is also produced in adult macrophages, brain cells, keratinocytes, and is widely expressed in cancer cells.
Precaución: For research use only.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty