Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tubastatin A hydrochloride ≥99% (by HPLC)


CHM240-0005 CHM240-0010 CHM240-0025 CHM240-0050 CHM240-0100 CHM240-0200 CHM240-0500
76203-422EA 125 USD InStock
76203-422 76203-424 76203-426 76203-428 76203-430 76203-432 76203-434
Tubastatin A hydrochloride ≥99% (by HPLC)
Tubastatin A hydrochloride
Tubastatin A hydrochloride is a potent and selective inhibitor of HDAC6. 

  • Potent and selective HDAC6 inhibitor

White solid.

Soluble in DMSO.


Formula: C₂₀H₂₁N₃O₂·HCl
Storage Temperature: Freezer
MDL Number: MFCD20488052
CAS Number: 1310693-92-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty