Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Carbon nanotube single-walled/double-walled ≥90%

Warning

06-0508-250MG 06-0508-1G
100197-308EA 128.8 USD InStock
100197-308 100210-958
Carbon nanotube single-walled/double-walled ≥90%
Carbon nanotube
Carbon Nanotubes, Nanomaterials

The tubes are 1 to 2 nm in diameter with lengths of 5 to 30 microns.
Ash is <1.5 wt%.


Formula: C
MDL Number: MFCD00133992
CAS Number: 308068-56-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty