CHM226-0001
CHM226-0005
CHM226-0010
CHM226-0025
76203-238EA
287
USD
InStock
76203-238
76203-240
76203-242
76203-244
Ixabepilone ≥99% (by HPLC) free base
Ixabepilone
Ixabepilone is an epothilone B analog and nontaxane microtubule-stabilizing compound with clinical activity in a range of solid tumors
- Nontaxane microtubule-stabilizing compound