PolyMag CRISPR, OZ Biosciences
Supplier: Med-Lab-Service
PNC40200 PNC41000 KPC40100
103537-328EA 364.62 USD InStock
103537-328 103537-330 103537-332
PolyMag CRISPR, OZ Biosciences
Transfection Reagents
PolyMag CRISPR kit is the only magnet assisted transfection reagent optimized to deliver high level of plasmid DNA and/or mRNA expressing Cas9 and guide RNA (gRNA).
  • High efficiency delivery of CRISPR/Cas9 system even on primary and hard to transfect cells
  • Ideal for co-transfection of pDNA/pDNA, pDNA/gRNA, pDNA/mRNA
  • Optimized for large plasmid DNA containing Cas9, and guide RNA
  • Low cell toxicity - enable to start your experiment with less cell
This reagent is based on our proprietary Magnetofection™ technology. The nanoparticles are composed of iron oxide magnetic core coated with cationic molecules. For your gene editing experiments, this reagent provides high transfection efficiency without toxicity.
Información de suministro: Shipped at room temperature.
Precaución: Store at +4 °C.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty