TC Diethrich Micro Needle Holder, OR Grade, Sklar®
Supplier: Sklar
21-5000 21-5001 21-5002 21-5003 21-5004
76192-576EA 1321.27 USD InStock
76192-576 76192-578 76192-580 76192-582 76192-584
TC Diethrich Micro Needle Holder, OR Grade, Sklar®
Forceps
The Sklar® Diethrich Micro Needle Holder is a finger ring instrument with a double spring movement and locking mechanism.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
The Sklar® Diethrich Micro Needle Holder has long, narrow shanks and very small jaws with cross-serrations used for holding micro size suture needles. Sklar® permanently bonds tungsten carbide inserts to the jaws, which maximizes control and precision when maneuvering the instrument and the needle.
Order Now
ORDER
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty