TC Ryder Micro Needle Holder, OR Grade, Sklar®
Supplier: Sklar
21-8015 21-8024
76192-652EA 545.09 USD InStock
76192-652 76192-654
TC Ryder Micro Needle Holder, OR Grade, Sklar®
Forceps
The Sklar® TC Ryder Micro Needle Holder is a ratcheted, finger ring instrument that features narrow jaws and is commonly used with very small suture needles in cardiovascular, plastic, and neurosurgical procedures.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
Tungsten carbide micro cross-serrated tips firmly grip the small needle and assist in keeping it from twisting during suturing.
Order Now
ORDER
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty