Carbon Steel Cylinders and Adapters, Strem
Supplier: Strem Chemicals Inc
96-8500-1EA 96-8100-1EA 96-8000-1CYL 96-8005-1CYL 96-8501-1EA 96-8105-1EA
100207-914EA 55.2 USD InStock
100207-914 100197-978 100209-242 100200-616 100207-916 100200-222
Carbon Steel Cylinders and Adapters, Strem
Cylinders Hydrometer Cylinders
This collection of carbon steel cylinders, valves, and adapters are designed primarily for the safe transport and storage of pyrophoric materials.
  • Type I Cylinder is a metal container fitted with a brass needle valve
  • Type II Cylinder is equipped with a brass ball valve
  • Cylinders are not equipped with a dip tube
The Type I Adapter consists of 6" brass tube (1/4" O.D.) connected to a 1/4" brass Swagelok® with a 1/4" female pipe thread. The Type II Adapter consists of 6" brass tube (1/4" O.D.) connected to a 1/4" brass Swagelok® with a 1/4" male pipe thread.
Precaución: As with any air sensitive compound, the cylinder should be taken into an inert atmosphere bag or box to remove the product.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty