Human Recombinant Annexin A7
Supplier: Novotek GmbH
C208-10UG C208-1MG C208-500UG C208-50UG
102860-470EA 118.46 USD InStock
102860-470 102860-472 102860-474 102860-476
Human Recombinant Annexin A7
Enzymes
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human Annexin A7 is produced by our E.coli expression system and the target gene encoding Met1-Gln466 is expressed.
Información accesorios: Accessories not included.
Información de suministro: Shipped Via FedEx
Envase: Shipped at ambient temperature via FedEx envelope.
Precaución: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty