CA38-10UG
CA38-50UG
CA38-500UG
CA38-1MG
102943-506EA
170.15
USD
InStock
102943-506
102943-508
102943-510
102943-512
Human Recombinant Linker for Activation of T-Cells Family Member 2
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
- Consistent Sizing options
- Datasheet Included
- High Purity, Low Endotoxin levels
Recombinant Human Non-T cell activation linker is produced by our Mammalian expression system and the target gene encoding Arg27-Ala243 is expressed with a 6His tag at the C-terminus.
Información accesorios: Accessories not included.
Información de suministro: Shipped Via FedEx
Envase: Shipped at ambient temperature via FedEx envelope.
Precaución: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.