Human Recombinant Interleukin-10 Receptor Subunit beta(IL10RB)
Supplier: Novotek GmbH
CC08-10UG CC08-1MG CC08-500UG CC08-50UG
102508-880EA 57.08 USD InStock
102508-880 102508-882 102508-884 102508-886
Human Recombinant Interleukin-10 Receptor Subunit beta(IL10RB)
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human IL-10 Receptor beta is produced by our Mammalian expression system and the target gene encoding Met20-Ser220 is expressed with a 6His tag at the C-terminus.
Información accesorios: Accessories not included.
Información de suministro: Shipped Via FedEx
Envase: Shipped at ambient temperature via FedEx envelope.
Precaución: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty