Human Recombinant Medium-Chain Specific Acyl-CoA Dehydrogenase Mitochondrial
Supplier: Novotek GmbH
CF46-10UG CF46-1MG CF46-500UG CF46-50UG
102864-790EA 118.46 USD InStock
102864-790 102864-792 102864-794 102864-796
Human Recombinant Medium-Chain Specific Acyl-CoA Dehydrogenase Mitochondrial
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human ACADM is produced by our E.coli expression system and the target gene encoding Lys26-Asn421 is expressed with a 6His tag at the N-terminus.
Información accesorios: Accessories not included.
Información de suministro: Shipped Via FedEx
Envase: Shipped domestic on polar packs in a 9"x6"x7" box, or shipped internationally on Dry Ice in a 14" cube box.
Precaución: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty