Human Recombinant Ube2B
Supplier: Novotek GmbH
CF58-10UG CF58-1MG CF58-500UG CF58-50UG
102864-878EA 118.46 USD InStock
102864-878 102864-880 102864-882 102864-884
Human Recombinant Ube2B
Enzymes
It offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products.
  • Consistent sizing options
  • Datasheet included
  • High purity, low endotoxin levels
Recombinant human ubiquitin-conjugating enzyme E2 B is produced by our E.coli expression system and the target gene encoding Met1-Ser152 is expressed.
Información accesorios: Accessories not included.
Información de suministro: Shipped via FedEx.
Envase: Shipped domestic on polar packs in a 9×6×7" box, or shipped internationally on dry ice in a 14" cube box.
Precaución: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only. The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty