Recombinant Mouse Cxcl16
Supplier: Novotek GmbH
CG20-10UG CG20-1MG CG20-500UG CG20-50UG
102865-230EA 34.46 USD InStock
102865-230 102865-232 102865-234 102865-236
Recombinant Mouse Cxcl16
Enzymes
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products.
  • Consistent sizing options
  • Datasheet included
  • High purity, low endotoxin levels
Recombinant Schistosoma Japonicum Glutathione S-transferase Class-mu 26 kDa Isozyme is produced by our E.coli expression system and the target gene encoding Met1-Lys218 is expressed.
Información accesorios: Accessories not included.
Información de suministro: Shipped Via FedEx.
Envase: Shipped at ambient temperature via FedEx envelope.
Precaución: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty