Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Hydroxyphenylboronic acid pinacol ester ≥97%

Warning

05-1042-1G 05-1042-5G
100199-700EA 151.45 USD InStock
100199-700 100199-702
2-Hydroxyphenylboronic acid pinacol ester ≥97%
2-Hydroxyphenylboronic acid pinacol ester
Boronic Acid and Ester


Formula: C₁₂H₁₇BO₃
Boiling Pt: 100 °C (0.2 mmHg)
MDL Number: MFCD02093754
CAS Number: 269409-97-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty