Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium cobalticarborane

Warning

05-2500-1G 05-2500-5G 05-2500-25G
100204-776EA 182.58 USD InStock
100204-776 100198-492 100207-202
Sodium cobalticarborane
Sodium cobalticarborane


MDL Number: MFCD00467949
CAS Number: 99492-72-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty