Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Carbon nanotube multi-walled, as produced cathode deposit

Warning

06-0504-5G 06-0504-1G
100197-304EA 274.58 USD InStock
100197-304 100210-954
Carbon nanotube multi-walled, as produced cathode deposit
Carbon nanotube
Carbon Nanotubes, Nanomaterials


Formula: C
MDL Number: MFCD00133992
CAS Number: 308068-56-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty