Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-[3,5-Bis(trifluoromethyl)phenyl]-3-[(1S,2S)-(+)-2-(dimethylamino)cyclohexyl]thiourea

Warning

07-0284-250MG 07-0284-1G
101433-068EA 66.52 USD InStock
101433-068 101433-070
1-[3,5-Bis(trifluoromethyl)phenyl]-3-[(1S,2S)-(+)-2-(dimethylamino)cyclohexyl]thiourea
1-[3,5-Bis(trifluoromethyl)phenyl]-3-[(1S,2S)-(+)-2-(dimethylamino)cyclohexyl]thiourea
Organocatalyst


Formula: C₁₇H₂₁F₆N₃S
MW: 413.4200 g/mol
Melting Pt: 87…88 °C
MDL Number: MFCD09834840
CAS Number: 851477-20-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty