Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Coproporphyrin I dihydrochloride (synthetic)


07-0300-50MG 07-0300-10MG
100201-342EA 457.17 USD InStock
100201-342 100201-340
Coproporphyrin I dihydrochloride (synthetic)
Coproporphyrin I dihydrochloride


MDL Number: MFCD00013469
CAS Number: 69477-27-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty