Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,3-Bis(2,6-diisopropylphenyl)imidazolidin-2-ylidene ≥98%

Danger

07-0593-500MG 07-0593-2G
101442-226EA 171.26 USD InStock
101442-226 101442-228
1,3-Bis(2,6-diisopropylphenyl)imidazolidin-2-ylidene ≥98%
1,3-Bis(2,6-diisopropylphenyl)imidazolidin-2-ylidene
NHC


Formula: C₂₇H₃₈N₂
MDL Number: MFCD10567379
CAS Number: 258278-28-3
UN: 1325
ADR: 4.1,II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty