Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Nitrilotriacetic acid disodium salt ≥99%


07-1475-250G 07-1475-50G
100202-734EA 325.54 USD InStock
100202-734 100204-808
Nitrilotriacetic acid disodium salt ≥99%
Nitrilotriacetic acid disodium salt


Formula: C₆H₇NNa₂O₆
MW: 235.1 g/mol
Melting Pt: > 300 °C
MDL Number: MFCD00064230
CAS Number: 15467-20-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty