Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-(+)-2,2'-Bis(methoxymethoxy)-1,1'-binaphthyl ≥98%

Warning

08-0170-5G 08-0170-250MG 08-0170-1G
100201-384EA 1166.28 USD InStock
100201-384 100201-380 100201-382
(R)-(+)-2,2'-Bis(methoxymethoxy)-1,1'-binaphthyl ≥98%
(R)-(+)-2,2'-Bis(methoxymethoxy)-1,1'-binaphthyl


Formula: C₂₄H₂₂O₄
MW: 374.429 g/mol
Melting Pt: 100…103 °C
MDL Number: MFCD03788935
CAS Number: 173831-50-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty