Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Perfluoro(2-butyltetrahydrofuran) ≥95%


09-5910-100G 09-5910-25G
100201-420EA 1174.77 USD InStock
100201-420 100196-012
Perfluoro(2-butyltetrahydrofuran) ≥95%
Perfluoro(2-butyltetrahydrofuran)
Organo Fluorine


Formula: C₈F₁₆O
Boiling Pt: 99…107 °C
Density: 1,77
MDL Number: MFCD00014513
CAS Number: 335-36-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty