Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6,6'-[(3,3'-Di-t-butyl-5,5'-dimethoxy-1,1'-biphenyl-2,2'-diyl)bis(oxy)] bis(dibenzo[d,f][1,3,2]dioxaphosphepin) hemi ethyl acetate adduct ≥95%

Warning

15-0107-1G 15-0107-250MG
101433-100EA 496.8 USD InStock
101433-100 101433-098
6,6'-[(3,3'-Di-t-butyl-5,5'-dimethoxy-1,1'-biphenyl-2,2'-diyl)bis(oxy)] bis(dibenzo[d,f][1,3,2]dioxaphosphepin) hemi ethyl acetate adduct ≥95%
6,6'-[(3,3'-Di-t-butyl-5,5'-dimethoxy-1,1'-biphenyl-2,2'-diyl)bis(oxy)] bis(dibenzo[d,f][1,3,2]dioxaphosphepin) hemi ethyl acetate adduct
Phosphine


Formula: C₄₆H₄₄O₈P₂
MW: 786.78 g/mol
Melting Pt: 188…190 °C
MDL Number: MFCD09839135
CAS Number: 121627-17-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty