Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

cis-1,2-Bis(diphenylphosphino)ethylene ≥98%

Warning

15-0250-5G 15-0250-1G
100210-792EA 441.6 USD InStock
100210-792 100197-394
cis-1,2-Bis(diphenylphosphino)ethylene ≥98%
cis-1,2-Bis(diphenylphosphino)ethylene
Phosphine


Formula: C₂₆H₂₂P₂
Melting Pt: 125…126 °C
MDL Number: MFCD00003046
CAS Number: 983-80-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty