Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-Phanephos ≥95%

Warning

15-0426-100MG 15-0426-500MG
100196-070EA 162.77 USD InStock
100196-070 100196-072
(S)-Phanephos ≥95%
(S)-Phanephos
PHANEPHOS, Phosphine


Formula: C₄₀H₃₄P₂
Melting Pt: 222…225 °C
MDL Number: MFCD03094575
CAS Number: 192463-40-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty