Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Cyclohexyldichlorophosphine ≥98%

Danger

15-0800-1G 15-0800-5G
100196-074EA 108.98 USD InStock
100196-074 100196-076
Cyclohexyldichlorophosphine ≥98%
Cyclohexyldichlorophosphine
Phosphine


Boiling Pt: 100 °C (17 mmHg)
MDL Number: MFCD00014295
CAS Number: 2844-89-5
UN: 1760
ADR: 8,II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty