Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-(+)-2-Diphenylphosphino-2'-methoxy-1,1'-binaphthyl ≥99%

Warning

15-1775-500MG 15-1775-100MG
100199-798EA 560.49 USD InStock
100199-798 100199-796
(R)-(+)-2-Diphenylphosphino-2'-methoxy-1,1'-binaphthyl ≥99%
(R)-(+)-2-Diphenylphosphino-2'-methoxy-1,1'-binaphthyl
Phosphine


Formula: C₃₃H₂₅OP
Melting Pt: 177…179 °C
MDL Number: MFCD00269686
CAS Number: 145964-33-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty