Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyldiphenylphosphine ≥99%

Warning

15-2150-5G 15-2150-25G
100200-430EA 86.34 USD InStock
100200-430 100200-428
Ethyldiphenylphosphine ≥99%
Ethyldiphenylphosphine
Phosphine


Formula: C₁₄H₁₅P
MDL Number: MFCD00015170
CAS Number: 607-01-2
UN: 3334
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty