Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Hexylphosphonic acid ≥97%

Warning

15-2410-1G 15-2410-5G
101418-650EA 75.02 USD InStock
101418-650 101442-422
Hexylphosphonic acid ≥97%
Hexylphosphonic acid


Formula: C₆H₁₅O₃P
MW: 166.16 g/mol
Melting Pt: 105…106 °C
MDL Number: MFCD00013961
CAS Number: 4721-24-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty