Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(11aS)-(-)-10,11,12,13-Tetrahydrodiindeno[7,1-de:1',7'-fg][1,3,2]dioxaphosphocin-5-dimethylamine ≥98%

Warning

15-5151-100MG 15-5151-500MG
101418-672EA 157.11 USD InStock
101418-672 101418-674
(11aS)-(-)-10,11,12,13-Tetrahydrodiindeno[7,1-de:1',7'-fg][1,3,2]dioxaphosphocin-5-dimethylamine ≥98%
(11aS)-(-)-10,11,12,13-Tetrahydrodiindeno[7,1-de:1',7'-fg][1,3,2]dioxaphosphocin-5-dimethylamine
MONOPHOS, Phosphine, SIPHOS


Formula: C₁₉H₂₀NO₂P
Melting Pt: 95…96 °C
MDL Number: MFCD08459340
CAS Number: 443965-10-4
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty