Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tri-i-butylphosphine ≥93%

Danger

15-5750-100G 15-5750-25G
100199-806EA 246.28 USD InStock
100199-806 100211-020
Tri-i-butylphosphine ≥93%
Tri-i-butylphosphine
Phosphine


Boiling Pt: 85 °C (7 mmHg)
MDL Number: MFCD00015049
CAS Number: 4125-25-1
UN: 3183
ADR: 4.2,II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty