Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tris[4-(trifluoromethyl)phenyl]phosphine ≥97%

Warning

15-8010-5G 15-8010-1G
100199-818EA 785.54 USD InStock
100199-818 100199-816
Tris[4-(trifluoromethyl)phenyl]phosphine ≥97%
Tris[4-(trifluoromethyl)phenyl]phosphine
Phosphine


Formula: C₂₁H₁₂F₉P
Melting Pt: 73…75 °C
MDL Number: MFCD00058883
CAS Number: 13406-29-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty