Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tris(trimethylsilyl)phosphine 10% (w/w) in hexanes ≥98%

Danger

15-8021-50G 15-8021-10G
100202-758EA 806.77 USD InStock
100202-758 100204-988
Tris(trimethylsilyl)phosphine 10% (w/w) in hexanes ≥98%
Tris(trimethylsilyl)phosphine
Phosphine


Formula: [(CH₃)₃Si]₃P
MDL Number: MFCD00015487
CAS Number: 15573-38-3
UN: 1993
ADR: 3,I
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty