Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Potassium triethylborohydride 1.0M in THF

Danger

Packed in a Sure/Seal™ bottle (Trademark of Sigma-Aldrich Co. LLC).

19-1970-100MMOL 19-1970-500MMOL
101418-754EA 485.48 USD InStock
101418-754 101418-756
Potassium triethylborohydride 1.0M in THF
Potassium triethylborohydride


MDL Number: MFCD00011702
CAS Number: 22560-21-0
UN: 3399
ADR: 4.3,I
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty