Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2,2,6,6-Tetramethyl-3,5-heptanedionato-O,O')potassium ≥95%


19-6000-5G 19-6000-25G 19-6000-1G
100203-026EA 318.46 USD InStock
100203-026 100199-228 100204-994
(2,2,6,6-Tetramethyl-3,5-heptanedionato-O,O')potassium ≥95%
(2,2,6,6-Tetramethyl-3,5-heptanedionato-O,O')potassium
Metal TMHD


Formula: C₁₁H₁₉KO₂
Boiling Pt: 150 °C (dec.)
MDL Number: MFCD00156522
CAS Number: 22441-14-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty