Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(4R,5R)-(-)-2,2-Dimethyl-α,α,α',α'-tetraphenyl-1,3-dioxolane-4,5-dimethanolato[1,2-bis(dimethoxy)ethane]titanium(IV) dichloride acetonitrile adduct

Warning

22-0780-250MG 22-0780-1G
100203-038EA 209.48 USD InStock
100203-038 100203-040
(4R,5R)-(-)-2,2-Dimethyl-α,α,α',α'-tetraphenyl-1,3-dioxolane-4,5-dimethanolato[1,2-bis(dimethoxy)ethane]titanium(IV) dichloride acetonitrile adduct
(4R,5R)-(-)-2,2-Dimethyl-α,α,α',α'-tetraphenyl-1,3-dioxolane-4,5-dimethanolato[1,2-bis(dimethoxy)ethane]titanium(IV) dichloride acetonitrile adduct
Stability: air sensitive, moisture sensitive


Formula: C₃₅H₃₈Cl₂O₆Ti
MDL Number: MFCD04973054
CAS Number: 328123-04-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty