Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Vanadium(IV) octaethylporphine oxide

Warning

23-5841-1G 23-5841-250MG
100208-856EA 1269.6 USD InStock
100208-856 100210-798
Vanadium(IV) octaethylporphine oxide
Vanadium(IV) octaethylporphine oxide
Porphine


Formula: C₃₆H₄₄N₄OV
MW: 601.718 g/mol
MDL Number: MFCD00058868
CAS Number: 27860-55-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty