Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Bis(cyclopentadienyl)manganese ≥98%

Danger

25-0200-5G 25-0200-25G 25-0200-1G
100208-864EA 519.45 USD InStock
100208-864 100197-450 100196-162
Bis(cyclopentadienyl)manganese ≥98%
Bis(cyclopentadienyl)manganese
Metallocenes, Derivatives & Cp Precursors, Volatile Organometallics for CVD & ALD, Volatile Precursors for CVD


Formula: C₁₀H₁₀Mn
Melting Pt: 172…173 °C
MDL Number: MFCD00045468
CAS Number: 73138-26-8
UN: 1325
ADR: 4.1,II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty