Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Bis(i-propylcyclopentadienyl)manganese ≥98%

Danger

25-0245-1G 25-0245-5G
100205-026EA 179.75 USD InStock
100205-026 100202-378
Bis(i-propylcyclopentadienyl)manganese ≥98%
Bis(i-propylcyclopentadienyl)manganese
Metallocenes, Derivatives & Cp Precursors, Volatile Organometallics for CVD & ALD, Volatile Precursors for CVD


MDL Number: MFCD01862453
CAS Number: 85594-02-1
UN: 1993
ADR: 3 II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty