Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,1'-Diethylferrocene ≥98%

Warning

26-0310-5G 26-0310-1G
100202-380EA 607.2 USD InStock
100202-380 100205-038
1,1'-Diethylferrocene ≥98%
1,1'-Diethylferrocene
Metallocenes, Derivatives & Cp Precursors, Volatile Organometallics for CVD & ALD, Volatile Precursors for CVD


Formula: C₁₄H₁₀Fe
Density: 1.18
MDL Number: MFCD01862455
CAS Number: 1273-97-8
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty