Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-(+)-1-[(R)-2-(Di-2-furylphosphino)ferrocenyl]ethyldi-3,5-xylylphosphine ≥97%

Warning

26-1170-100MG 26-1170-500MG
100209-832EA 157.11 USD InStock
100209-832 100204-580
(S)-(+)-1-[(R)-2-(Di-2-furylphosphino)ferrocenyl]ethyldi-3,5-xylylphosphine ≥97%
(S)-(+)-1-[(R)-2-(Di-2-furylphosphino)ferrocenyl]ethyldi-3,5-xylylphosphine
JOSIPHOS, Phosphine


Formula: C₃₆H₃₆FeO₂P₂
MW: 618.4700 g/mol
MDL Number: MFCD06658128
CAS Number: 649559-66-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty